Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Difluoroacetic acid, 98%

Catalog No. AAA1278706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1278706 5 g
AAA1278714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1278706 Supplier Thermo Scientific Chemicals Supplier No. A1278706
Add to Cart

CAS: 381-73-7 | C2H2F2O2 | 96.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-73-7
Molecular Formula C2H2F2O2
Molecular Weight (g/mol) 96.033
MDL Number MFCD00004220
InChI Key PBWZKZYHONABLN-UHFFFAOYSA-N
Synonym difluoroacetic acid, acetic acid, difluoro, difluoressigsaeure, 1,1-difluoroacetic acid, unii-zqk1c95k3n, zqk1c95k3n, acetic acid, 2,2-difluoro, difluoroaceticacid, difluoracetic acid, difluoro-acetic acid
PubChem CID 9788
ChEBI CHEBI:23716
IUPAC Name 2,2-difluoroacetic acid
SMILES C(C(=O)O)(F)F

Specifications

Melting Point -1°C
Density 1.526
Boiling Point 132°C to 134°C
Flash Point 78°C (172°F)
Odor Pungent
Refractive Index 1.343
Quantity 5 g
UN Number UN3265
Beilstein 1098588
Formula Weight 96.03
Percent Purity 98%
Chemical Name or Material Difluoroacetic acid
Show More Show Less
missing translation for 'healthHazard1' H227-H314
missing translation for 'healthHazard2' GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
missing translation for 'healthHazard3' P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-839-0
RTECSNumber AG9900000
TSCA Yes

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.