Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Difluoromethyl Phenyl Sulfone

Catalog No. 74260010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-260-010G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 74-260-010G Supplier Sigma Aldrich Supplier No. 74260010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent Used forReductive silylation and the preparation of trifluoro- and difluoromethylsilanes by reductive coupling of fluoromethyl sulfones, sulfoxides and sulfides with chlorosilanesFluoroalkylation/chloroalkylation of +a,+a-enones, arynes, acetylenic ketones and other Michael acceptorsDifluoromethylation of primary alkyl halides via nucleophilic substitution-reductive desulfonylationReagent used in Preparation of+a-difluoromethyl amines via stereoselective (phenylsulfonyl)difluoromethylation of chiral sulfinyl aldiminesAnti-difluoropropanediols via potassium tert-butoxide-catalyzed difluoromethylenation of aldehydes+a-difluoromethylated and +a-difluoromethylenated alcohols and amines by regioselective nucleophilic difluoromethylation of 1,2-cyclic sulfates and sulfamidatesDifluoroalkenes from alkyl halides via nucleophilic substitution-eliminationDifluoromethyl alcohol derivatives from enolizable and non-enolizable carbonyl compounds using nucleophilic phenylsulfonyldifluoromethylation-reductive desulfonylation strategyFluoromethylated vicinal ethylenediamines via fluoromethylation of chiral +a-aminobutanesulfinimines with (phenylsulfonyl)fluoromethanes followed by reductive desulfonylation and alcoholysis

Specifications

CAS 141037-05-0
Density 1.092 g/mL at 25°C
Linear Formula C8H10N2O2
Quantity 500 mg
Synonym Nano particles, magnetic, IDA functionalized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.