Learn More
Cayman Chemical Difluprednate, Purity: ≥98%, CAS: 23674-86-4, 50 mg
SDP
Catalog No. 502995060
Encompass_Preferred
A synthetic glucocorticoid (Ki = 78 pM for the glucocorticoid receptor); increases intraocular pressure in sheep following ocular administration of a 0.05% w/v solution
Catalog No. 50-299-5060 Supplier Cayman Chemical Supplier No. 2377650 MG
A synthetic glucocorticoid (Ki = 78 pM for the glucocorticoid receptor); increases intraocular pressure in sheep following ocular administration of a 0.05% w/v solution