Learn More
AdipoGen DiFMUEt
SDP
Supplier:  AdipoGen CDXD0164M050
Encompass_Preferred
Chemical. CAS 215868-24-9. Formula C12H10F2O3. MW 240.2. Synthetic. Non-fluorescent, fluorogenic cytochrome P450 substrate. Converted by ethoxyresorufin O-deethylase ECOD to the blue fluorescent product DiFMU CDX-D0059.
Catalog No. 50-256-1206