Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Difopein TFA | 96.30% | 6387.17 (free acid) | 5 MG
SDP

Catalog No. 5000423244
Encompass_Preferred

Difopein TFA is a specific and competitive inhibitor of the 14-3-3 protein, a highly conserved eukaryotic regulatory molecule. It functions by blocking the ability of 14-3-3 to bind to target proteins and inhibits 14-3-3/Ligand interactions. This compound is not cell permeable and induces apoptosis, enhancing the ability of Cisplatin to kill cells.

  • Specific and competitive inhibitor of 14-3-3 protein
  • Blocks 14-3-3 from binding to target proteins
  • Inhibits 14-3-3/ligand interactions
  • Induces apoptosis
  • Enhances cisplatin's ability to kill cells
  • Target: 14-3-3 protein
  • Soluble in DMSO at ≥ 100 mg/mL
  • In vivo soluble at ≥ 2.5 mg/mL with specific protocol
  • Recommended storage for powder: -80°C for 2 years; -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-23244 Supplier Medchemexpress LLC Supplier No. HYP1380A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Difopein TFA is a specific and competitive inhibitor of the 14-3-3 protein, a highly conserved eukaryotic regulatory molecule. It functions by blocking the ability of 14-3-3 to bind to target proteins and inhibits 14-3-3/Ligand interactions. This compound is not cell permeable and induces apoptosis, enhancing the ability of Cisplatin to kill cells.

  • Specific and competitive inhibitor of 14-3-3 protein
  • Blocks 14-3-3 from binding to target proteins
  • Inhibits 14-3-3/ligand interactions
  • Induces apoptosis
  • Enhances cisplatin's ability to kill cells
  • Target: 14-3-3 protein
  • Soluble in DMSO at ≥ 100 mg/mL
  • In vivo soluble at ≥ 2.5 mg/mL with specific protocol
  • Recommended storage for powder: -80°C for 2 years; -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.