Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Digital Control Metering Pump, 18.9 L/hr; 115 VAC, 60 Hz
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7630422
Encompass
Digital units offer pinpoint accuracy. They feature an LED readout, 4 to 50 mA, and a 4000:1 turndown ratio. The variable units offer the same digital control with the additional variable speed control capability.
Catalog No. NC1486079