Learn More
Chemglass Life Sciences Digital Controller Only, for Brushless DC Motor, Closed Loop Controls, TFT Display, Wifi, 115V-230V
SDP
Catalog No. 501941130
Encompass_Preferred
Digital Controller Only, for Brushless DC Motor, Closed Loop Controls, TFT Display, Wifi, 115V-230V. Velp/Chemglass Wi-Fi Enabled Laboratory overhead stirrer motor and separate controller. Controller features a TFT 3.5” Display that shows all setting information at a glance. Wi-Fi Connection to Velp Ermes Platform allows the end user monitor status and data logging on the cloud. USB interface for use with optional PC software to data log, monitor, turn off/on. USB also allow Software Updates. Stirrerfeatures torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Features: Timer For Unattended Operations, Compatible with CG-2033-B 1/4 HP Brushless Motor, Key Lock Function to Avoid Unintended Changes with Settings, Programmable methods to setup to 4 Time/Speed Ramps, PT100 Probe Connection for Temperature Monitoring: Measurement Range -200 to +550° C
Catalog No. 50-194-1130 Supplier Chemglass Life Sciences Supplier No. CG2033V75
Digital Controller Only, for Brushless DC Motor, Closed Loop Controls, TFT Display, Wifi, 115V-230V. Velp/Chemglass Wi-Fi Enabled Laboratory overhead stirrer motor and separate controller. Controller features a TFT 3.5” Display that shows all setting information at a glance. Wi-Fi Connection to Velp Ermes Platform allows the end user monitor status and data logging on the cloud. USB interface for use with optional PC software to data log, monitor, turn off/on. USB also allow Software Updates. Stirrerfeatures torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Features: Timer For Unattended Operations, Compatible with CG-2033-B 1/4 HP Brushless Motor, Key Lock Function to Avoid Unintended Changes with Settings, Programmable methods to setup to 4 Time/Speed Ramps, PT100 Probe Connection for Temperature Monitoring: Measurement Range -200 to +550° C