Learn More
Research Products International Corp Digital Disruptor Genie
SDP
Catalog No. 502229783
Encompass
Digital cell disruptor with complete control of speed and time. Ideal for releasing biological molecules from inside a cell. More than vortexing alone, patented multi-directional action simultaneously agitates and vortexes at high-speed producing greater yields in less time.
Supplied with Universal Microtube Holder (tubes not included) and Pop-off Cup.
Catalog No. 50-222-9783 Supplier Research Products International Corp Supplier No. 155598
Digital cell disruptor with complete control of speed and time. Ideal for releasing biological molecules from inside a cell. More than vortexing alone, patented multi-directional action simultaneously agitates and vortexes at high-speed producing greater yields in less time.
Supplied with Universal Microtube Holder (tubes not included) and Pop-off Cup.