Learn More
Biotang Inc Digital Dry Bath (block) incubator with cooling, a microprocessor-controlled product, more stable and accurate temperature control is achieved. Temp. -10deg.C to 100deg.C, temp. Control Accuracy: 0.3,11 blocks selective.
Catalog No. 507514971 Shop All BIOTANG Inc Products
Encompass_Preferred
Digital Dry Bath (block) incubator with cooling, a microprocessor-controlled product, more stable and accurate temperature control is achieved. Temp. -10deg.C to 100deg.C, temp. Control Accuracy: 0.3,11 blocks selective.
Catalog No. 50-751-4971 Supplier Biotang Inc Supplier No. MK20
Digital Dry Bath (block) incubator with cooling, a microprocessor-controlled product, more stable and accurate temperature control is achieved. Temp. -10deg.C to 100deg.C, temp. Control Accuracy: 0.3,11 blocks selective.