Learn More
Biotang Inc Digital Dry Bath (block) incubator a microprocessor-controlled product, more stable and accurate temperature control is achieved. Temp. 5deg.C above ambient to 100deg.C, temp. Control Accuracy: 0.3,11 blocks selective.
Catalog No. 507514970 Shop All BIOTANG Inc Products
Encompass_Preferred
Digital Dry Bath (block) incubator a microprocessor-controlled product, more stable and accurate temperature control is achieved. Temp. 5deg.C above ambient to 100deg.C, temp. Control Accuracy: 0.3,11 blocks selective.
Catalog No. 50-751-4970 Supplier Biotang Inc Supplier No. MK10
Digital Dry Bath (block) incubator a microprocessor-controlled product, more stable and accurate temperature control is achieved. Temp. 5deg.C above ambient to 100deg.C, temp. Control Accuracy: 0.3,11 blocks selective.