Learn More
Biotium Digoxigenin-dUTP, alkali stable
Catalog No. 501964657
Encompass_Preferred
Digoxigenin-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3′-end terminal labeling to synthesize labeled DNA probes for in-situ hybridization, microarray or blotting techniques. The digoxigenin labeled probe could be detected with a fluorescent labeled or enzyme labeled anti-digoxigenin antibody.
Catalog No. 50-196-4657 Supplier Biotium Supplier No. 40078
Digoxigenin-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3′-end terminal labeling to synthesize labeled DNA probes for in-situ hybridization, microarray or blotting techniques. The digoxigenin labeled probe could be detected with a fluorescent labeled or enzyme labeled anti-digoxigenin antibody.