Learn More
Thermco Products Inc DIG FRZR THERMTR CERT @78C
Catalog No. NC0859921
Encompass_Preferred
Ultra Low Freezers Digital Thermometer is able to monitor temperatures down to -200°C and to -100°C with buffer medium bottle. Thermometer is NIST Traceable and is supplied with a certificate. The probe is housed in a 60ml PETG plastic bottle, filled with USP sand which insulates the sensitive section of the probe to maintain temperature during the opening and closing of the door.• NIST Traceable• Range: -200 to 1376°C (-328 to 2498°F) Note: without bottle• Range: -100 to 160°C (-148 to 320°F) Note: with bottle• Resolution: 0.1°C• Accuracy: 0.3% +1.0°C• Celsius & Fahrenheit• Dimensions: 160mm x 54mm x 34mm• Power: 1 9v Battery• Min-Max Function• Temperature Hold Function• Auto Shut Off After 30 mins• CE Approved
Catalog No. NC0859921 Supplier Thermco Products Inc Supplier No. ACCULF307
Ultra Low Freezers Digital Thermometer is able to monitor temperatures down to -200°C and to -100°C with buffer medium bottle. Thermometer is NIST Traceable and is supplied with a certificate. The probe is housed in a 60ml PETG plastic bottle, filled with USP sand which insulates the sensitive section of the probe to maintain temperature during the opening and closing of the door.• NIST Traceable• Range: -200 to 1376°C (-328 to 2498°F) Note: without bottle• Range: -100 to 160°C (-148 to 320°F) Note: with bottle• Resolution: 0.1°C• Accuracy: 0.3% +1.0°C• Celsius & Fahrenheit• Dimensions: 160mm x 54mm x 34mm• Power: 1 9v Battery• Min-Max Function• Temperature Hold Function• Auto Shut Off After 30 mins• CE Approved