Learn More
MSC Igaging EZ Absolute Digital Height Gauge, Stainless Steel, 0-8 in. (200 mm), 1 ea
Catalog No. 5000512281
Encompass
The EZ Absolute Digital Height Gauge provides precise vertical measurements with a range of 0 to 200 mm (0 to 8 in.). Designed for reliable workshop use, this tool features a durable stainless steel beam and an iP54 rating for resistance against dust and moisture. The integrated absolute encoding technology retains the origin zero position even when the gauge is powered off, ensuring consistent measurement accuracy.
  • Carbide tipped scriber for enhanced durability
  • Large LCD display for clear readouts
  • Zero setting and inch/metric conversion available at any position
  • Micro USB data output port for connectivity
  • Single stainless steel beam with lightweight construction
  • Fine adjustment mechanism for ultra-smooth movement
  • Supplied in a protective molded case
  • Auto power off to preserve battery life
Catalog No. 50-005-12281 Supplier MSC Supplier No. 44506186
The EZ Absolute Digital Height Gauge provides precise vertical measurements with a range of 0 to 200 mm (0 to 8 in.). Designed for reliable workshop use, this tool features a durable stainless steel beam and an iP54 rating for resistance against dust and moisture. The integrated absolute encoding technology retains the origin zero position even when the gauge is powered off, ensuring consistent measurement accuracy.
  • Carbide tipped scriber for enhanced durability
  • Large LCD display for clear readouts
  • Zero setting and inch/metric conversion available at any position
  • Micro USB data output port for connectivity
  • Single stainless steel beam with lightweight construction
  • Fine adjustment mechanism for ultra-smooth movement
  • Supplied in a protective molded case
  • Auto power off to preserve battery life