Learn More
Hach Company Dig. Incer. 3/4in ORP Sen-PVDF
Catalog No. 5000208090 Shop All Hach Company Products
Encompass
Digital 3/4 Inch ORP Sensor-Insertion Sensor Style-PVDF Body Material-Platinum Electrode-Temperature Compensation - NonePVDF Insertion Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable
Catalog No. 50-002-08090 Supplier Hach Company Supplier No. DRC2K5N
Digital 3/4 Inch ORP Sensor-Insertion Sensor Style-PVDF Body Material-Platinum Electrode-Temperature Compensation - NonePVDF Insertion Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable