Learn More
Research Products International Corp Digital Incubator Shaker, Mini Shaker, CO2, Compact
SDP
Catalog No. 502229543
Encompass
The Incu-Shaker CO2 Mini is the most compact shaking CO2 incubator in its class. Ideal shaker for culturing nonadherent cells with combined precision and orbital shaking for optimum conditions for cell culture. Unique and proprietary heat distribution technology allows the incubator to provide the highest levels of temperature accuracy with minimal fluctuation (within 0.1°C). The six-side heating system in combination with a low speed internal fan uniformly spreads air throughout the stainless steel chamber. Beltless motor drive provides a horizontally circular shaking motion at a consistent 19mm orbit. The included platform (9.5 x 11.5 in.) can be used for low speed shaking of a variety of laboratory vessels, including Erlenmeyer flasks, culture plates and media bottles. For high speed mixing, a patented MAGic Clamp platform is available that allows for magnetic attachment of flask clamps and tube racks.
Catalog No. 50-222-9543 Supplier Research Products International Corp Supplier No. 248571
Add to Cart
Add to Cart
Request a Quote
The Incu-Shaker CO2 Mini is the most compact shaking CO2 incubator in its class. Ideal shaker for culturing nonadherent cells with combined precision and orbital shaking for optimum conditions for cell culture. Unique and proprietary heat distribution technology allows the incubator to provide the highest levels of temperature accuracy with minimal fluctuation (within 0.1°C). The six-side heating system in combination with a low speed internal fan uniformly spreads air throughout the stainless steel chamber. Beltless motor drive provides a horizontally circular shaking motion at a consistent 19mm orbit. The included platform (9.5 x 11.5 in.) can be used for low speed shaking of a variety of laboratory vessels, including Erlenmeyer flasks, culture plates and media bottles. For high speed mixing, a patented MAGic Clamp platform is available that allows for magnetic attachment of flask clamps and tube racks.