Learn More
Bellco Glass, Inc. Digital Mini-Orbital Shaker Complete, 115V
SDP
Catalog No. 501945965
Encompass_Preferred
The Bellco Digital Mini-Orbital Shaker is a high speed shaker for general laboratory cell culture experimentation. This Bellco design incorporates an orbital platform that accepts a variety of optional trays. Peg and grommet design offers a simple method to install or change trays; four versions of trays are available. Brushless DC motor designed for minimal maintenance. MADE IN THE USA
Catalog No. 50-194-5965 Supplier Bellco Glass, Inc. Supplier No. 764420115
The Bellco Digital Mini-Orbital Shaker is a high speed shaker for general laboratory cell culture experimentation. This Bellco design incorporates an orbital platform that accepts a variety of optional trays. Peg and grommet design offers a simple method to install or change trays; four versions of trays are available. Brushless DC motor designed for minimal maintenance. MADE IN THE USA