Learn More
Research Products International Corp Digital Multi-MicroPlate Genie Pulse
SDP
Supplier:  Research Products International Corp 155581
Encompass
High speed mixing action with up to 4 microplates, or 8 with optional accessory tray. Speed and orbit combine to offer true vortexing action in each well of the microplate. Configurable speed and time for mixing consistency. Ideal for applications such as small volume mixing, PCR Preparation, Digestions, as well as Cell Disruption.
Catalog No. 50-222-9821