Learn More
Sigma Aldrich Fine Chemicals Biosciences DIG Nick Translation Mix
Catalog No. 502905470
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.A The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.
Catalog No. 50-290-5470 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11745816910
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.A The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.