Learn More
Sigma Aldrich Fine Chemicals Biosciences DIG RNA Labeling Mix, 40 uL
Catalog No. NC1883959
Encompass
RNA labeling with Digoxigenin-11-UTP by in vitro transcription with SP6, T7, and T3 RNA polymerases. DIG-labeled RNA is used in a variety of hybridization techniques.
Catalog No. NC1883959 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11277073910
RNA labeling with Digoxigenin-11-UTP by in vitro transcription with SP6, T7, and T3 RNA polymerases. DIG-labeled RNA is used in a variety of hybridization techniques.