Learn More
Cole-Parmer Cole-Parmer Stuart Digital Stirring Hot Plate, Ceramic, White; 120 VAC
Catalog No. 501960397 Shop All Cole-Parmer Products
Encompass
These custom designed stirring hot plates provide accurate stirring and temperature control. They feature a hot surface warning light that flashes when the plate temperature is above 50C. Its durable front panel resists spills and chemicals. The plates are available in various interface, plate surface,and body color configurations.
Catalog No. 50-196-0397 Supplier Cole-Parmer Supplier No. 0480644
These custom designed stirring hot plates provide accurate stirring and temperature control. They feature a hot surface warning light that flashes when the plate temperature is above 50C. Its durable front panel resists spills and chemicals. The plates are available in various interface, plate surface,and body color configurations.