Learn More
Cole-Parmer Cole-Parmer Stuart Digital Magnetic Stirrer with Timer, 20L Capacity, 110V
Catalog No. 501960389 Shop All Cole-Parmer Products
Encompass_Preferred
Sleek low-profile digital stirrer has a bright LCD showing both set and working parameters. Unique control pad allows all operations to be set using just your fingertip. Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built-in timer can be set for up to 9 hours or continuous operation.
Catalog No. 50-196-0389 Supplier Cole-Parmer Supplier No. 0466129
Sleek low-profile digital stirrer has a bright LCD showing both set and working parameters. Unique control pad allows all operations to be set using just your fingertip. Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built-in timer can be set for up to 9 hours or continuous operation.