Learn More
Thermco Products Inc DIG WP THERMOMTER 50-300C
Catalog No. NC1285470
Encompass_Preferred
-50/300C & -58/572F, Waterproof, 8" Stem, Cert @ +/- 0.5C
Catalog No. NC1285470 Supplier Thermco Products Inc Supplier No. ACC6558SUP
-50/300C & -58/572F, Waterproof, 8" Stem, Cert @ +/- 0.5C