Learn More
Medchemexpress LLC HY-U00145 1mg Medchemexpress, Digeranyl bisphosphonate CAS:878143-03-4 Purity:>98%
SDP
Catalog No. 501934426
Encompass_Preferred
Medchemexpress, HY-U00145 1mg Digeranyl bisphosphonate CAS:878143-03-4 Digeranyl bisphosphonate is a potent geranylgeranylpyrophosphate (GGPP) synthase inhibitor, which inhibits geranylgeranylation of Rac1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4426 Supplier Medchemexpress LLC Supplier No. HYU001451MG
Medchemexpress, HY-U00145 1mg Digeranyl bisphosphonate CAS:878143-03-4 Digeranyl bisphosphonate is a potent geranylgeranylpyrophosphate (GGPP) synthase inhibitor, which inhibits geranylgeranylation of Rac1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.