Learn More
Invitrogen™ Digest-All 4 (Proteinase K)
Catalog No. 003011
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
00-301-1 50 mL
1 options
Catalog No. 00-301-1 Supplier Invitrogen™ Supplier No. 003011
Add to Cart
Edge
Add to Cart
This product may be used to pretreat formalin-fixed, paraffin-embedded tissues (see Invitrogen Anatomical Pathology catalog page 140), and will improve the signal of certain antigens.
This product may be used to pretreat formalin-fixed, paraffin-embedded tissues (see Invitrogen Anatomical Pathology catalog page 140), and will improve the signal of certain antigens. It contains 50 mL of ready-to-use Proteinase K. Digest-All™ 4 is stable for 12 months.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 mL