Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digestion Acid, Water : Nitric Acid : Hydrochloric Acid, 2 : 2 : 1, Ricca Chemical
SDP

Catalog No. RIC01000SOL Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RIC01000SOL 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RIC01000SOL Supplier Ricca Chemical Company Supplier No. 25535PT
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Digestion Acid
Color Yellow
CAS 7697-37-2, 7732-18-5, 7647-01-0
CAS Min % 48.0, 31.69, 18.7
CAS Max % 50.0, 32.98, 19.46
Grade Laboratory
Concentration or Composition (by Analyte or Components) Water:Nitric Acid Hydrochloric Acid, 2:2:1
Packaging Safety-coated Glass Bottle
pH <1
Quantity 2.5 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.