Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digestion Reagent, with Copper Catalyst, Ricca Chemical
SDP

Catalog No. R2551000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2551000500 500 mL
2551-32 1 L
2551-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R2551000500 Supplier Ricca Chemical Company Supplier No. 255116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Analysis of Nitrogen (Kjeldahl)

Specifications

Chemical Name or Material Digestion Reagent
Name Note With Copper Catalyst
CAS Min % 0.51, 23.95
Color Blue
CAS Max % 8.81, 0.53
CAS 7778-80-5, 7664-93-9, 7732-18-5, 7758-99-8
Physical Form Liquid
Quantity 500 mL
Packaging Natural Poly Bottle
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.