Learn More
SCP Science DIGESTION TUBES 750/CS
Catalog No. NC9739574
Encompass
DigiTUBE without RackLock, Features: Specially Selected Polypropylene , Leak Free Screw Cap , Accurate 50ml Graduation, Frosted Writing Area, Transparent
Catalog No. NC9739574 Supplier SCP Science Supplier No. 010500263
DigiTUBE without RackLock, Features: Specially Selected Polypropylene , Leak Free Screw Cap , Accurate 50ml Graduation, Frosted Writing Area, Transparent