Learn More
Azzota Corp Digital Honey Test Refractometer, Nd: 1.4419-1.5232, Be: 33.1-47.7, Water: 38.4%-5.4%, Power Supply: 1Battery (9V)
Catalog No. 501612827
Encompass_Preferred
Digital Honey Test Refractometer, Nd: 1.4419-1.5232, Be: 33.1-47.7, WATER: 38.4%-5.4%, Power Supply: 1battery (9v)
Catalog No. 50-161-2827 Supplier Azzota Corp Supplier No. DRH300
Digital Honey Test Refractometer, Nd: 1.4419-1.5232, Be: 33.1-47.7, WATER: 38.4%-5.4%, Power Supply: 1battery (9v)