Learn More
Cole-Parmer Digi-Sense 1-Zone Temperature Controller; Ramp/Soak, Type J, 120V/12A
Supplier:  Cole-Parmer 3622566
Encompass_Preferred
These controllers are designed for controlling multiple steps in a predetermined temperature processes, and features modern control technology that produces greater temperature stability. The Ramp/Soak feature of this control allows up to a 40-step profile, which can be configured from the provided software. You can profile temperature, time, hold, soak and end steps to create the ideal profile for your process. Auto-tune minimizes setpoint overshoot and features the ability to learn your process producing greater stability. The built in adaptive control technology provides even tighter control for these demanding applications. The controller also features an audible alarm function, temperature selectable and one panel mounted receptacle that accept a three-prong plug. Select from models for types J, K, or T thermocouples with miniconnectors, or 100 ohm platinum RTD probes. This unit comes with a lattice support bracket, which is ideal for mounting to maximize space.
Catalog No. NC1659807