Learn More
Cole-Parmer Traceable Big-Digit Thermocouple Thermometer with Calibration
Catalog No. NC1272855 Shop All Cole-Parmer Products
Encompass_Preferred
Thermometer reads from 58 to 2000F and 50 to 1300C with a resolution of 0.1 and 1. Switch from F to C at the touch of a button. Easy to read big digits measure over 5/8-inch-high and are readable from 15 feet away. Unit features HOLD button that freezes display to capture current reading. MAX HOLD feature permits capturing and displaying the highest temperature achieved over any time period.
Catalog No. NC1272855 Supplier Cole-Parmer Supplier No. 9121007
Thermometer reads from 58 to 2000F and 50 to 1300C with a resolution of 0.1 and 1. Switch from F to C at the touch of a button. Easy to read big digits measure over 5/8-inch-high and are readable from 15 feet away. Unit features HOLD button that freezes display to capture current reading. MAX HOLD feature permits capturing and displaying the highest temperature achieved over any time period.