Learn More
Cole-Parmer Digi-Sense Dry Block/Incubator Liquid-In-Glass Thermometer; 0 to 100C, 35mm Immersion, Organic Liquid Fill
Catalog No. NC1244381 Shop All Cole-Parmer Products
Encompass_Preferred
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PFA coated for added safety which may cause a slight change in accuracy up to 1 division. Dry block thermometers - designed for use in all standard heating blocks and water baths with most having arrows indicating critical temperatures.
Catalog No. NC1244381 Supplier Cole-Parmer Supplier No. 0800616
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PFA coated for added safety which may cause a slight change in accuracy up to 1 division. Dry block thermometers - designed for use in all standard heating blocks and water baths with most having arrows indicating critical temperatures.