Learn More
Cole-Parmer Traceable 2-Input Data Logging Thermocouple Probe Thermometer, Type K/J
Supplier:  Cole-Parmer 2025002
Encompass_Preferred
Handheld thermocouple thermometers provide accurate and reliable temperature measurement in an economic and compact design. Features include Min/Max/Avg, Hold, °F/°C or Kelvin (K) selectable units, relative time stamp on Min/Max/Avg, electronic off set function and auto power-off to save on battery power. Meters accept both type K or J thermocouple probes with a miniconnector. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA.  It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC0869803