Learn More
Cole-Parmer RTD Probe, 100 Ohm, Digi-Sense 3-Pin Connector, 4" Length, 5 ft Coil Cord
Catalog No. NC1641195 Shop All Cole-Parmer Products
Encompass_Preferred
Designed to give the same accuracy and benefits of a standard RTD probe in a compact economical design. Time Constant: 3 seconds or greater. Custom designs or changes to this product are available upon request.
Catalog No. NC1641195 Supplier Cole-Parmer Supplier No. 9382100
Designed to give the same accuracy and benefits of a standard RTD probe in a compact economical design. Time Constant: 3 seconds or greater. Custom designs or changes to this product are available upon request.