Learn More
Cole-Parmer DIGI SENSE TC MOUNTING PANEL
Supplier:  Cole-Parmer 1852743
Encompass_Preferred
Digi-Sense Thermocouple Mounting Panel, Vertical, Standard Connectors; 5 Circuits. Use to creat easy to access direct temperature connection in control rooms, test stands, laboratories, and plants. Use with insert panel temperature connectors to create a complete panel assembly. Available in 2, 4, 5 & 6 circuits for Vertical Face Plates and 6, 8, 10 & 12 circuits for Horizontal Face Plates.
Catalog No. NC2008483