Learn More
Cole-Parmer Traceable Temperature/RH Touch Screen Recorder with NIST-Calibration
Catalog No. NC1294581 Shop All Cole-Parmer Products
Encompass_Preferred
Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA.  It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC1294581 Supplier Cole-Parmer Supplier No. 2025040
Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA.  It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.