Learn More
Cole-Parmer Digi-Sense Calibrated Liquid-In-Glass Thermometer; 0 to 300F, 76mm Immersion, Organic Liquid Fill
Catalog No. NC1474139 Shop All Cole-Parmer Products
Encompass_Preferred
These thermometers are ideal for use in applications where a very precise accurate temperature measurement is required. Thermometers are individually calibrated to NIST standards at specific temperatures and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division. Thermometers feature triangular anti-roll fitting, permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an NIST-traceable statement of accuracy and a triangular-style antiroll fitting.
Catalog No. NC1474139 Supplier Cole-Parmer Supplier No. 9030020
These thermometers are ideal for use in applications where a very precise accurate temperature measurement is required. Thermometers are individually calibrated to NIST standards at specific temperatures and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division. Thermometers feature triangular anti-roll fitting, permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an NIST-traceable statement of accuracy and a triangular-style antiroll fitting.