Learn More
Cole-Parmer Traceable Ultra-Low Temperature Wi-Fi Data Logger with TraceableLIVE Cloud Service; 1 Probe
Catalog No. NC1754427 Shop All Cole-Parmer Products
Encompass_Preferred
Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability. View current conditions anywhere-cloud-based interface sends email, text, and push notifications.
Catalog No. NC1754427 Supplier Cole-Parmer Supplier No. 1800027
Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability. View current conditions anywhere-cloud-based interface sends email, text, and push notifications.