Learn More
Cole-Parmer Digi-Sense Ultra Low Liquid-In-Glass Thermometer; -100 to 50C, Total Immersion
Catalog No. NC1050960 Shop All Cole-Parmer Products
Encompass_Preferred
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.
Catalog No. NC1050960 Supplier Cole-Parmer Supplier No. 9030028
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.