Learn More
SCP Science DigiTUBE, 100ml, NRL, 300 per Pack
Catalog No. NC0198149
Encompass
Non-RackLock DigiTUBEs, graduated every 10mls along with readings at 25, 50, 75 & 100ml levels. Class 'A' calibrated at the 50 and 100ml mark. Complete with caps made of virgin polypropylene with the lowest leachable metal available. Note: For use with 100ml DigiPREP systems.
Catalog No. NC0198149 Supplier SCP Science Supplier No. 010501263
Non-RackLock DigiTUBEs, graduated every 10mls along with readings at 25, 50, 75 & 100ml levels. Class 'A' calibrated at the 50 and 100ml mark. Complete with caps made of virgin polypropylene with the lowest leachable metal available. Note: For use with 100ml DigiPREP systems.