Learn More
SCP Science DigiTUBEs 50ml RackLock no caps, 750/cs
Catalog No. NC9977501
Encompass
Manufactured using virgin polypropylene, verified for leachable metals by ICP-MS. Low leachable catalytic/additive metal content, excellent visibility and good acid resistance are guaranteed. Tubes are graduated, in 5 ml increments, up to the 50 ml graduation mark. The 50 ml graduation mark is certified accurate to ±0.25 ml in accordance with ASTM Class "A" volumetric standards. This reduces and eliminates the need for graduated cylinders and volumetric flasks.
Catalog No. NC9977501 Supplier SCP Science Supplier No. 010500262
Manufactured using virgin polypropylene, verified for leachable metals by ICP-MS. Low leachable catalytic/additive metal content, excellent visibility and good acid resistance are guaranteed. Tubes are graduated, in 5 ml increments, up to the 50 ml graduation mark. The 50 ml graduation mark is certified accurate to ±0.25 ml in accordance with ASTM Class "A" volumetric standards. This reduces and eliminates the need for graduated cylinders and volumetric flasks.