Learn More
SCP Science SCP Science DigiTUBE 50 mL, RackLock Design, Leak-Free Blue Caps, HandyPAK Packaging, 750 Tubes
Catalog No. NC9295926
Encompass
The SCP Science DigiTUBE 50 mL RackLock HandyPAK is a versatile solution for analytical laboratories, offering a high-performance design for sample preparation, storage, and digestion. Made of virgin polypropylene with ultra-low leachable metals, these tubes ensure reliable performance at temperatures below 130°C. Their innovative RackLock feature allows secure placement in racks and provides easy, leak-free access with screw caps. With 750 tubes per case, they effectively replace multiple traditional lab vessels, reducing costs and saving time.
Catalog No. NC9295926 Supplier SCP Science Supplier No. 010500362
The SCP Science DigiTUBE 50 mL RackLock HandyPAK is a versatile solution for analytical laboratories, offering a high-performance design for sample preparation, storage, and digestion. Made of virgin polypropylene with ultra-low leachable metals, these tubes ensure reliable performance at temperatures below 130°C. Their innovative RackLock feature allows secure placement in racks and provides easy, leak-free access with screw caps. With 750 tubes per case, they effectively replace multiple traditional lab vessels, reducing costs and saving time.