Learn More
SCP Science SCP Science DigiTUBE 50 mL, Low Leachable Metal Content, Accurate to ±0.25 mL, HandyPAK Packaging
Catalog No. NC1817749
Encompass
The SCP Science DigiTUBE 50 mL tubes offer exceptional sample integrity and precision for analytical labs. Made from virgin polypropylene, they exhibit low leachable metal content, ensuring accurate results. Designed for digestion below 130°C, these graduated tubes streamline lab processes by replacing multiple traditional vessels, improving efficiency and reducing costs. Packaged in HandyPAK with leak-free blue screw caps, they provide durability and cleanliness during use. Ideal for various digestion and analytical applications.
Catalog No. NC1817749 Supplier SCP Science Supplier No. 010500364
The SCP Science DigiTUBE 50 mL tubes offer exceptional sample integrity and precision for analytical labs. Made from virgin polypropylene, they exhibit low leachable metal content, ensuring accurate results. Designed for digestion below 130°C, these graduated tubes streamline lab processes by replacing multiple traditional vessels, improving efficiency and reducing costs. Packaged in HandyPAK with leak-free blue screw caps, they provide durability and cleanliness during use. Ideal for various digestion and analytical applications.