Learn More
Perkin Elmer US LLC DigiFILTER 1.0 micron for 50 mL DigiTUBEs
Supplier:  Perkin Elmer US LLC N9308032
Encompass_Preferred
The vacuum assisted DigiFILTER assembly provides a quick and easy way to filter samples prior to analysis. Increase lab productivity by filtering particulates from digested samples in a matter of seconds directly from the digestion tube.
Catalog No. 50-210-3747