Learn More
Electron Microscopy Sciences Digimatic Caliper Absolute Function with Spc Output
SDP
Catalog No. 50996489
Encompass_Preferred
Provides a digital readout to 0.0005" / 0.01 mm for error-free reading and can measure steps. Function: zero setting, data hold, low battery alarm and data output. Provided with a data saving output port for connection with the SPC system. Powered by a single SR-44 battery. Battery life: approximately two years under normal use. Specifications: An all metal head with a plastic housing. Range: 0 – 6" / 0 - 150 mm; LCD resolution: 0.0005" / 0.0; Accuracy: +/-0.001
Catalog No. 50-996-489 Supplier Electron Microscopy Sciences Supplier No. 6206220
Provides a digital readout to 0.0005" / 0.01 mm for error-free reading and can measure steps. Function: zero setting, data hold, low battery alarm and data output. Provided with a data saving output port for connection with the SPC system. Powered by a single SR-44 battery. Battery life: approximately two years under normal use. Specifications: An all metal head with a plastic housing. Range: 0 – 6" / 0 - 150 mm; LCD resolution: 0.0005" / 0.0; Accuracy: +/-0.001