Learn More
Cole-Parmer Traceable Remote Probe Thermometer with Calibration; ±1.0°C accuracy (-20 to 100°C)
Catalog No. NC1556464 Shop All Cole-Parmer Products
Encompass_Preferred
Magnetic back allows for placement on metal surfaces such as freezers, incubators, or water baths. Includes a flip-open stand for use on bench or desktop, and a spring fastener for clipping onto any edge. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC1556464 Supplier Cole-Parmer Supplier No. 9020520
Magnetic back allows for placement on metal surfaces such as freezers, incubators, or water baths. Includes a flip-open stand for use on bench or desktop, and a spring fastener for clipping onto any edge. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.