Learn More
Cole-Parmer Traceable Excursion-Trac™ Data Logging Thermometer with Calibration; 1 Bottle Probe
Catalog No. NC1650186 Shop All Cole-Parmer Products
Encompass_Preferred
Rolling memory retains most recent 525,600 temperature observations
Catalog No. NC1650186 Supplier Cole-Parmer Supplier No. 9446007
Rolling memory retains most recent 525,600 temperature observations