Learn More
Hach Company Digit. Conv. 3/4in ORP Sen-PPS
Catalog No. 5000208089 Shop All Hach Company Products
Encompass
Digital 3/4 Inch ORP Sensor-Convertible Sensor Style-PPS Body-Platinum Electrode-Temperature Compensation-NonePPS Convertible Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable
Catalog No. 50-002-08089 Supplier Hach Company Supplier No. DRC1R5N
Digital 3/4 Inch ORP Sensor-Convertible Sensor Style-PPS Body-Platinum Electrode-Temperature Compensation-NonePPS Convertible Body Style 4 5m Analog Cable Platinum Electrode No Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable