Learn More
Cole-Parmer Dri-Block Digital Heater, triple insert, 200C; 115 V
Supplier:  Cole-Parmer 3662011
Encompass
Techne Dri-Block Digital Block Heater, Triple Insert, 200C; 115 VAC, Precision-engineered heaters with excellent temperature accuracyVersatile range of interchangeable heating blocks work with many size sample tubes and platesTake up less storage spacelightweight and flat-top surface allow units to be stacked when not in use115 VAC operation,INCLUDESBlock extraction tool and a power cord with US plug.
Catalog No. NC0907421