Learn More
Motic™ Swift Line Digital 6.0 MP Microscope Camera
SCI_ED
Catalog No. S81725
Change view
Click to view available options
Resolution:
6.0 MP
Catalog No. Resolution
S81725 6.0 MP
1 options
Catalog No. S81725 Supplier Motic™ Swift Line Supplier No. DMOTICAM S6
Add to Cart
Add to Cart
Easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
Motic's new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. Easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
Included Motic Images software enables live and captured images at a maximum quality. Edit, measure, and share images or reports with this comprehensive software.
INCLUDES
  • CS ring
  • Mini USB 3.1 cable (removable)
  • Motic 4-dot Calibration slide
  • Motic Images Plus 3.0 for PC and Mac
CAMERA SPECIFICATIONS
  • 6.0 MP 1/1.8 in. sCMOS
  • USB 3.1
  • 3072 x 2048 @ 30 fps
  • 1536 x 1024 @ 50 fps
REQUIRED SPECIFICATIONS
  • Processor: 2 Ghz multi-core or above
  • USB 2.0 (Minimum Requirement)
  • USB 3.0 (Required for high speed transmission)
  • RAM: 8 GB
  • Hard Disk: 5 GB (Free Space)
  • Video Card: MUST have dedicated 1 GB Video RAM
Specifications
Type USB Microscope Camera
Resolution 6.0 MP
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov