Learn More
Azzota Corp Digital Abbe Refractometer, Refractive Index Measurement Range: 1.3000-1.7000 Nd, Dissolved Solids Measurement Range: 0-95% Brix; Refractive Index Accuracy: <0.0002 Nd
Catalog No. 507532957
Encompass_Preferred
Digital Abbe Refractometer, Refractive index measurement range: 1.3000-1.7000 nD, Dissolved solids measurement range: 0-95% Brix; Refractive index accuracy: <0.0002 nD
Catalog No. 50-753-2957 Supplier Azzota Corp Supplier No. AR3
Digital Abbe Refractometer, Refractive index measurement range: 1.3000-1.7000 nD, Dissolved solids measurement range: 0-95% Brix; Refractive index accuracy: <0.0002 nD